Urea, high purity
Supplier: VWR
Synonyms:
Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
0568-12KG
0568-1KG
0568-2.5KG
0568-500G
0568-5KG
0568-10KG
97061-918EA
1478.88
CAD
97061-918
97061-920
97061-922
97061-924
97061-914
97061-926
97061-916
Urea, high purity
Urea
Commonly used to denature nucleic acids for electrophoresis and to study the secondary and tertiary structure of proteins. Effective for maintaining DNA in a highly denatured state during sequencing protocols, urea is also useful for accelerating the activity of Proteinase K digestions. Effectively denatures RNA. For use in nucleic electrophoresis applications.
Formula:
NH₂CONH₂ Boiling Pt: 197 °C (1013 hPa) Melting Pt: 132.7 °C Density: 1.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008022 CAS Number: 57-13-6 EINECS: 200-315-5 REACH: 01-2119463277-33 Merck Index: 13,09935 |
|
Specification Test Results
Abs.@280nm (5M, Water) |
|
Biuret Test | Pass |
Chloride |
|
Cyanate |
|
DNase | none detected |
Heavy Metals (as Pb) |
|
Insolubles |
|
Iron |
|
Melting Point | 132°C - 135°C |
Protease | none detected |
Purity |
|
RNase | none detected |
Solubility (5M, Water) | Pass |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...