Diethyl pyrocarbonate ≥97.5%, high purity

Supplier: VWR
Warning

E174-100G E174-25G
97062-652EA 48.66 CAD
97062-652 97062-648 97062-650
Diethyl pyrocarbonate ≥97.5%, high purity
Diethyl pyrocarbonate
Formula: O(COOC₂H₅)₂
Boiling Pt: 93…94 °C
Density: 1.123 g/cm³ (20 °C)
Flash Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088

Order Now

Specification Test Results

Purity (acid assay) 97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR