Polyethylene glycol 8,000 for biotechnology
Supplier: VWR
|
Warning
|
Synonyms:
PEG 8,000, Ethylene glycol polymer 8000, PEG 8000
0159-2.5KG
0159-1KG
0159-500G
97061-100EA
313.61
CAD
97061-100
97061-098
97061-102
Polyethylene glycol 8,000 for biotechnology
Polyethylene glycol 8,000
Polyethylene glycol is used for the isolation of plasmid DNA and the precipitation of phage.
Formula:
H(OCH₂CH₂)nOH MW: 8000 g/mol |
MDL Number:
MFCD01779615 CAS Number: 25322-68-3 |
Specification Test Results
DNase | none detected |
pH (5%, Water) @25°C | 4.5 - 7.5 |
RNase | none detected |
Solidification Point | 55 °C - 64 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...