Dextran sulfate sodium salt, M.W. ~ 500,000, Ultra Pure Grade

Supplier: VWR
Warning

Synonyms: Dextran sulfate sodium from Dextran, Dextran sulphate sodium from Dextran

0198-250G 0198-50G
97061-194CS 11142.57 CAD
97061-194 97061-196
Dextran sulfate sodium salt, M.W. ~ 500,000, Ultra Pure Grade
Dextran sulphate sodium salt
Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Refrigerator
MDL Number: MFCD00081551
CAS Number: 9011-18-1

Order Now

Specification Test Results

DNase none detected
Heavy Metals = 0.005 %
RNase none detected
Iron = 0.001 %
Loss on Drying = 10 %
pH (1%, Water) @25°C 6.4 - 8
Protease none
Purity 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR