Polyvinylpyrrolidone, M.W. ∼ 40,000, high purity (K30)

Supplier: VWR

0507-500G 0507-1KG
97061-820CS 632.21 CAD
97061-820 97061-818
Polyvinylpyrrolidone, M.W. ∼ 40,000, high purity (K30)
Polyvinylpyrrolidone

Common reagent for nucleic acid hybridization procedures for detection of DNA/RNA.

Formula: (C₆H₉NO)n
MDL Number: MFCD01076626
CAS Number: 9003-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7893e4d0affb7fb76c948d932b4a0e880dece26c&vid=7893e4d0affb7fb76c948d932b4a0e880dece26c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Heavy Metals 0.001 %
pH (5%, Water) @25°C 3 - 7
Purity (Dry Basis) 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR