VWR® DNA Molecular Weight Markers, 100 bp Ladder
Supplier: VWR
N856-300UL
K180-250UL
97063-350EA
264.36
CAD
97063-350
97063-208
97063-488
VWR® DNA Molecular Weight Markers, 100 bp Ladder
Electrophoresis Reagents
Molecular Weight Markers
13 fragments ranging from 100 to 3000 bp for easy band identification.
- Easy band identification
- Available with loading dye
Also available pre-mixed with loading dye or with EZ-Vision® One DNA dye.