VWR® SDS-Page Gel Buffers
Supplier: Avantor
VWR®
VWRVM197-500ML
97063-954EA 354.69 CAD
97063-954
VWR® SDS-Page Gel Buffers
Buffers Electrophoresis Buffers Running Buffers
The 4X concentrated buffer (0.5M Tris pH 6.8 and 0.4% SDS) for preparation of stacking gels for Laemmli SDS-Page
  • Reduce pipetting steps with convenient 4X solutions
Dilution to a 1X solution results in final 1X concentrations of 0.125M Tris and 0.1% SDS .