VWR® Neutral pH Polyacrylamide Gel Electrophoresis Buffers
Supplier: VWR
K855-500ML
K856-500ML
97063-670EA
195.83
CAD
97063-670
97063-672
VWR® Neutral pH Polyacrylamide Gel Electrophoresis Buffers
Buffers
Electrophoresis Buffers
Running Buffers
Tris-Acetate-SDS, MOPS-SDS, and MES-SDS are useful for high resolution of proteins on neutral pH SDS-PAGE gels.
- Ultra-Pure Grade
- Convenient concentrations and packaging
- Excellent quality, consistency and resolution