D-(+)-Sucrose 20% in aqueous solution, Proteomics Grade

Supplier: VWR

Synonyms: α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranosideSucroseD(+)-Saccharoseβ-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranosideα-ᴅ-Glc-(1→2)-β-ᴅ-Fru

VWRVE543-100ML
97062-874EA 187.86 CAD
97062-874
D-(+)-Sucrose 20% in aqueous solution, Proteomics Grade
D(+)-Sucrose

Common reagent in routine nucleic acid procedures and electrophoresis loading dyes.

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
MDL Number: MFCD00006626
CAS Number: 57-50-1
https://ca.vwr.com/stibo/web/std.lang.all/95/86/2299586.jpg

Order Now

Specification Test Results

Clarity Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR