VWR® TRIS Buffer, 1 M Solution, pH 8.0

Supplier: VWR

E199-100ML E199-500ML
97062-672EA 101.97 CAD
97062-672 97062-674
VWR® TRIS Buffer, 1 M Solution, pH 8.0
Buffers Cell Culture Buffers

Tris (hydroxymethyl) aminomethane


  • Biotechnology grade
  • pH @25C 7.9 - 8.1


Storage conditions: Room temperature

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR