Sodium dodecyl sulfate (SDS) 20% in aqueous solution, Biotechnology Grade
Supplier: VWR
|
Warning
|
0837-200ML
0837-500ML
97062-440EA
140
CAD
97062-440
97062-442
Sodium dodecyl sulfate (SDS) 20% in aqueous solution, Biotechnology Grade
Sodium dodecyl sulphate (SDS)
Formula:
CH₃(CH₂)₁₁OSO₃Na Density: 1…1.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 |
|
Specification Test Results
Conductivity | 22800 umhos - 27600 umhos |
pH @25°C | 5 - 8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...