VWR® Serafree™ Cryopreservation Media
Supplier: VWR
N676-50ML N655-50ML
97063-278EA 460.25 CAD
97063-278 97064-276
VWR® Serafree™ Cryopreservation Media
Cell Freezing Media
SeraFree™ Cryopreservation Media is a ready-to-use freezing media for cryopreservation of adherent or suspension cultured cells.
  • Endotoxin tested
  • Reduces potential for transmission of infectious agents
  • Eliminates regulatory compliance requirements associated with use of animal products
  • Cost effective
The animal-free RPMI or DMEM based media composition eliminates batch-to-batch variability and are optimized for cell viability and cell growth after thawing.