VWR® Premixed LB Broth
Supplier: VWR
J106-2KG J106-1KG J106-500G
97064-112EA 531.54 CAD
97064-112 97064-110 97064-114
VWR® Premixed LB Broth
Media for Microbiology Dehydrated Media for Microbiology
Lysogeny broth (LB) is a nutritionally rich medium used for the growth of bacteria.
  • Save Time Using Premixed and Ready-to-Use Media
  • Homogenous and High Quality Powders
Lysogeny broth (LB) is a nutritionally rich medium used for the growth of bacteria. LB continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli. Powdered LB is available in premixed Miller or Lennox formulations and in Ready-Pack™ pouches which prepare 1L of media.
Formulation of 10 g tryptone, 10 g NaCl, and 5 g yeast extract (25 g powder total) per L of liquid broth.
The instructions included on the VWRVJ106 label state to prepare LB broth by dissolving 25 g powder per liter in deionized water.