VWR® SOB Broth, Biotechnology Grade
Supplier: Avantor
VWR®
VWRVJ906-500G J906-100G
97063-454CS 1859.61 CAD
97063-452 97063-454
VWR® SOB Broth, Biotechnology Grade
Media for Microbiology Dehydrated Media for Microbiology
SOB broth powder is used for the preparation of nutritionally rich growth medium and transformation of competent <italic>E
SOB Broth (Super Optimal Broth) growth medium is nutritionally rich, containing casein hydrolysate and yeast extract as sources of nitrogenous compounds and growth factors,and magnesium sulfate as a supply of magnesium ions necessary for DNA replication.
when dissolved at 25.5 g per liter of water,yields the formulation
Tryptone -------- 20 Grams/L
Yeast Extract- - -5 Grams/L
Sodium Chloride--0.5 Gram/L
Medium should be autoclaved or sterile-filtered. Allow the solution to cool to 50 °C or below before adding supplemental components and antibiotic. Add 20 mL of 1 M MgSO4.Store SOB Broth powder at room temperature (18 – 26 °C). Store prepared medium cold (2 – 8 °C).
coli.</italic>