Sodium chloride 5 M in aqueous solution, Biotechnology Grade

Supplier: VWR

Synonyms: HaliteSea saltNaClSaline

VWRVE529-500ML
97062-858EA 113.46 CAD
97062-858
Sodium chloride 5 M in aqueous solution, Biotechnology Grade
Sodium chloride

Common source of salts for media propagation of many organisms

Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Specification Test Results

Conductivity (1%, Water) 5000 - 7000 umhos
Dnase NONE / Fail
Identification PASS / Fail
pH @25C 5.0 - 9.0
Rnase NONE / Fail

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR