D-(+)-Glucose for biotechnology

Supplier: VWR

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

0188-50KG 0188-500G 0188-12KG 0188-1KG 0188-2.5KG 0188-5KG
97061-170EA 2883.85 CAD
97061-170 97061-168 97061-162 97061-164 97061-166 97061-172
D-(+)-Glucose for biotechnology
D(+)-Glucose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Arsenic = 0.00004%
Chloride = 0.01%
DNase none detected
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.005%
Iron = 0.0005%
Loss on Drying = 0.2%
Protease none detected
Purity = 99.5%
Residue on Ignition = 0.02%
RNase none detected
Specific Rotation 52.5°- 54 °
Sulfate & Sulfite = 0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR