D-(-)-Fructose, powder, high purity
Supplier: VWR
Synonyms:
D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
0226-5KG
0226-12KG
0226-1KG
97061-242EA
381.38
CAD
97061-242
97061-234
97061-236
97061-238
D-(-)-Fructose, powder, high purity
D(-)-Fructose
Fine white powder.
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Boiling Pt: 552 Melting Pt: 100…110 °C Density: 1.59 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00148910 CAS Number: 57-48-7 EINECS: 200-333-3 Merck Index: 13,04295 |
Specification Test Results
Arsenic |
|
Calcium & Magnesium |
|
Chloride |
|
Heavy Metals (as Pb) |
|
Loss on Drying |
|
Purity |
|
Residue on Ignition |
|
Sulfate |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...