VWR® Ultra Pure Grade Phosphate Buffered Saline
Supplier: VWR
K812-500ML
K812-20L
97063-658EA
129.1
CAD
97063-658
97063-656
VWR® Ultra Pure Grade Phosphate Buffered Saline
Buffers
Cell Culture Buffers
1X Solution Composition:137 mM NaCl, 2.7 mM KCl, 9.5 mM Phosphate buffer
- Does not contain magnesium or calcium.
- Solution is 0.2 µm filtered into bottles and autoclaved.
- Ready-to-use as supplied.
- pH at 25°C (1X PBS): 7.1 – 7.7
- DNase, RNase, Protease none detected.
- Suitable for cell culture.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...