Glycine Biotechnology

Supplier: VWR

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

0167-10KG 0167-1KG 0167-50KG 0167-12KG 0167-5KG
97061-124EA 1189.91 CAD
97061-124 97061-128 97061-130 97061-126 97061-132
Glycine Biotechnology
Glycine
Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Boiling Pt: 181
Melting Pt: 83
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@260 nm (1M solution) ≤0.1
Abs.@280 nm (1M solution) ≤0.1
Chloride = 0.01%
DNase none detected
Heavy Metals (as Pb) = 0.002%
Loss on Drying = 0.2%
pH (5%, Water) @25°C 5.9 - 6.4
Protease none detected
Purity = 99%
RNase none detected
Solubility (10%, Water) Pass
Sulfate = 0.05%
Thin Layer Chromatography (TLC) One Spot


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR