L(+)-Asparagine, anhydrous, high purity

Supplier: VWR

Synonyms: H-Asn-OH(S)-(+)-AsparagineL-Asparagine

VWRV94341-100G
97062-610CS 2113.57 CAD
97062-610
L(+)-Asparagine, anhydrous, high purity
L(+)-Asparagine
Formula: H₂NCOCH₂CH(NH₂)CO₂H
MW: 132.12 g/mol
MDL Number: MFCD00064401
CAS Number: 70-47-3

Order Now

Specification Test Results

Chloride (%) 0.05
Moisture (%) 0.5
Purity (dried basis) (%) 99.0
Sulfate (%) 0.05

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR