Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
Synonyms:
Methyl sulfoxide
, DMSO
, Dimethyl(oxido)sulfur
, Methyl sulphoxide
, Methanesulphinylmethane
, Dimethyl(oxido)sulphur
, Methanesulfinylmethane
VWRVN182-5X10ML
97063-136EA
290.94
CAD
97063-136
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Dimethyl sulphoxide
Formula:
(CH₃)₂SO MW: 78.14 g/mol Boiling Pt: 189 °C (1013 hPa) Melting Pt: 18.5 °C Density: 1.101 g/cm³ (20 °C) Flash Pt: 87 °C |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 Merck Index: 13,03285 |
Specification Test Results
Acidity Test | Pass |
Congealing Point |
|
Appearance | Clear colorless liquid. |
Non-volatile Residue |
|
Product Lock Test | C9481 |
Purity |
|
Refractive Index | 1.4755 - 1.4775 |
Specific Gravity | 1.095 - 1.101 |
Water (KF) | < = 0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...