VWR® TRIS Glycine Buffer, 10X Liquid Concentrate
Supplier: VWR
0307-4L
0307-1L
97061-380EA
254.63
CAD
97061-380
97061-378
VWR® TRIS Glycine Buffer, 10X Liquid Concentrate
Buffers
Electrophoresis Buffers
Running Buffers
A 1X working solution contains 0.025M Tris base and 0.192 M Glycine.
- Convenient concentrations and packaging
pH (1X, Water) @25°C - 8.3 - 8.7