Methanol ≥99.9%, BAKER ANALYZED ULTRA LC/MS™ for LC-MS, tested for UHPLC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = N
BAKR9863-02
CAJT9863-1CS
1144.46
CAD
CAJT9863-1
CAJT9863-2
Methanol ≥99.9%, BAKER ANALYZED ULTRA LC/MS™ for LC-MS, tested for UHPLC, J.T.Baker®
Methanol
ULTRA LC/MS & UHPLC
Formula:
CH₃OH Boiling Pt: 64.6 °C (1013 hPa) Melting Pt: –98 °C Density: 0.7918 g/cm³ (20 °C) Flash Pt: 11 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II REACH: 01-2119433307-44 Merck Index: 13,05984 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Appearance | Passes Test |
Gradient Elution Test (a.u.) - 254 nm | ≤0.01 |
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission | ≤0.3 ppb |
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities | ≤1.0 ppb |
Assay (CH₃OH) | ≥99.9 % |
Color (APHA) | ≤10 |
Residue after Evaporation | ≤1.0 ppm |
Water (H₂O) | ≤500 ppm |
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) | ≤25.0 ppb |
LC/MS Suitability - Largest Response on ESI- Negative Mode (as 4-Nitrophenol) | ≤25.0 ppb |
Trace Impurities - Aluminum (Al) | ≤10.0 ppb |
Trace Impurities - Calcium (Ca) | ≤20 ppb |
Trace Impurities - Chromium (Cr) | ≤5.0 ppb |
Trace Impurities - Cobalt (Co) | ≤5.0 ppb |
Trace Impurities - Copper (Cu) | ≤5.0 ppb |
Trace Impurities - Iron (Fe) | ≤5.0 ppb |
Trace Impurities - Lead (Pb) | ≤5.0 ppb |
Trace Impurities - Lithium (Li) | ≤10.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤10.0 ppb |
Trace Impurities - Manganese (Mn) | ≤5.0 ppb |
Trace Impurities - Nickel (Ni) | ≤5.0 ppb |
Trace Impurities - Potassium (K) | ≤10.0 ppb |
Trace Impurities - Sodium (Na) | ≤50.0 ppb |
Trace Impurities - Tin (Sn) | ≤5.0 ppb |
Trace Impurities - Zinc (Zn) | ≤10.0 ppb |
Filtered through a 0.1 micron filter. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...