Methanol ≥99.8%, Biograde for DNA and peptides synthesis

Supplier: Thermo Scientific Chemicals
Danger

44571-K7 44571-M6
CAAAA44571-K7EA 143.94 CAD
CAAAA44571-K7 CAAAA44571-M6
Methanol ≥99.8%, Biograde for DNA and peptides synthesis
Methanol

In nucleic acid and peptide synthesis


Purity: 99.8+% CAS No.: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04 Flash Point: 11°(52°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3290 Odor: Mild Melting Point: -98° Boiling Point: 64.7° Beilstein Reference: 1098229 Merck Reference: 14,5957 UN No.: 1230 MDL No.: MFCD00004595


In nucleic acid and peptide synthesis

Formula: CH₃OH
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.791
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 14,5957
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR