Dichloromethane, anhydrous ≥99.7% stabilised, ChemSeal™
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with amylene
Packaged under argon in resealable ChemSeal™ bottles.
41835-AK
41835-K2
41835-M1
CAAA41835-AKEA
66.2
CAD
CAAA41835-AK
CAAA41835-K2
CAAA41835-M1
Dichloromethane, anhydrous ≥99.7% stabilised, ChemSeal™
Dichloromethane
Packaged under Argon. Stabilized with amylene
Solvent for anhydrous reactions, water <0.01%.
Solvent for anhydrous reactions, water
Formula:
CH₂Cl₂ Boiling Pt: 39…40 °C Melting Pt: –95 °C Density: 1.325 Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1,III Merck Index: 14,6063 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...