2-Propanol ≥99.7% for gas chromatography
Supplier: Thermo Scientific Chemicals
|
Danger
|
40983-K2
40983-K7
40983-M1
CAAA40983-K2EA
96.29
CAD
CAAA40983-K2
CAAA40983-K7
CAAA40983-M1
2-Propanol ≥99.7% for gas chromatography
2-Propanol
Suitable for environmental, pesticide, and GC analysis
Purity: 99.7+%
CAS No.: 67-63-0
Molecular Formula: (CH3)2CHOH
Formula Weight: 60.10
Flash Point: 12°(53°F)
Physical Form: Liquid
Density: 0.785
Refractive Index: 1.3770
Odor: Alcohol-like
Melting Point: -90°
Boiling Point: 82-83°
Beilstein Reference: 635639
Merck Reference: 14,5208
UN No.: 1219
MDL No.: MFCD00011674
Suitable for environmental, pesticide, and GC analysis
Formula:
(CH₃)₂CHOH Boiling Pt: 82…83 °C Melting Pt: –90 °C Density: 0.785 Flash Pt: 12 °C (53 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II REACH: 01-2119457558-25 Merck Index: 14,5208 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...