Dichloromethane ≥99.5% stabilised ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methylene chloride
Stabilization:
Stabilised with amylene
39116-AP
39116-K2
39116-K4
39116-K7
39116-M1
CAAA39116-APEA
62.88
CAD
CAAA39116-AP
CAAA39116-K2
CAAA39116-K4
CAAA39116-K7
CAAA39116-M1
Dichloromethane ≥99.5% stabilised ACS
Dichloromethane
Stabilized with amylene
Purity: 99.5+%
CAS No.: 75-09-2
Molecular Formula: CH2Cl2
Formula Weight: 84.93
Physical Form: Liquid
Density: 1.325
Refractive Index: 1.4244
Appearance: Colorless
Odor: Sweet
Melting Point: -95°
Boiling Point: 39-40°
Beilstein Reference: 1730800
Merck Reference: 14,6063
UN No.: 1593
MDL No.: MFCD00000881
Formula:
CH₂Cl₂ Boiling Pt: 39…40 °C Melting Pt: –95 °C Density: 1.325 Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1,III Merck Index: 14,6063 |
|
Specification Test Results
Color (APHA) | 10 |
Evaporation residue | 0.00002 |
Titratable acid | 0.0003meq/g |
Free halogens | Pass test |
H2O | 0.0002 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...