Dichloromethane ≥99.7% stabilised for spectrophotometry
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methylene chloride
Stabilization:
Stabilised with amylene
32440-K2
32440-K7
32440-M1
CAAA32440-K2EA
113.51
CAD
CAAA32440-K2
CAAA32440-K7
CAAA32440-M1
Dichloromethane ≥99.7% stabilised for spectrophotometry
Dichloromethane
Stabilized with amylene
Purity: 99.7+%
CAS No.: 75-09-2
Molecular Formula: CH2Cl2
Formula Weight: 84.93
Physical Form: Liquid
Density: 1.325
Refractive Index: 1.4244
Odor: Sweet
Melting Point: -95°
Boiling Point: 39-40°
Beilstein Reference: 1730800
Merck Reference: 14,6063
UN No.: 1593
MDL No.: MFCD00000881
Formula:
CH₂Cl₂ MW: 84.93 g/mol Boiling Pt: 39.8 Melting Pt: -95 Density: 1.325 Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1 III Merck Index: 14,6063 |
|
Specification Test Results
Evaporation residue | 3 ppm |
H2O | 0.0002 |
Free halogens | Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...