Methanol absolute ≥99.8% ACS low acetone
Supplier: Thermo Scientific Chemicals
|
Danger
|
31721-AP
31721-K2
31721-K7
31721-M1
CAAA31721-APEA
49.45
CAD
CAAA31721-AP
CAAA31721-K2
CAAA31721-K7
CAAA31721-M1
Methanol absolute ≥99.8% ACS low acetone
Methanol
MDL: MFCD00004595
Beilstein Registry No.: 1098229
Purity: 99.8+%
CAS No.: 67-56-1
Molecular Formula: CH3OH
Formula Weight: 32.04
Flash Point: 11°(52°F)
Physical Form: Liquid
Density: 0.791
Refractive Index: 1.3290
Odor: Mild
Melting Point: -98°
Boiling Point: 64.7°
Beilstein Reference: 1098229
Merck Reference: 14,5957
UN No.: 1230
MDL No.: MFCD00004595
Formula:
CH₃OH Boiling Pt: 64.7 °C Melting Pt: –98 °C Density: 0.791 Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II REACH: 01-2119433307-44 Merck Index: 14,5957 |
|
Specification Test Results
Appearance | Clear |
Color (APHA) | 10 |
H2O | 0.1% |
Evaporation residue | 0.001% |
Solubility in water | Pass test |
Carbonyl compounds | 0.001 % each of acetone formaldehyde and acetaldehyde |
Titratable acid | 0.0003meq/g |
Titratable base | 0.0002meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...