Dichloromethane ≥99.7% stabilised for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methylene chloride
Stabilization:
Stabilised with amylene
MDL: MFCD00000881
Beilstein Registry No.: 1730800
Notes: Filtered through 0.2µ filters.
Fieser: 1,676 2,273 4,337 7,239 18,131
Purity: 99.7+%
CAS No.: 75-09-2
Molecular Formula: CH2Cl2
Formula Weight: 84.93
Physical Form: Liquid
Density: 1.325
Refractive Index: 1.4244
Odor: Sweet
Melting Point: -95°
Boiling Point: 39-40°
Beilstein Reference: 1730800
Merck Reference: 14,6063
UN No.: 1593
MDL No.: MFCD00000881
Formula:
CH₂Cl₂ MW: 84.93 g/mol Boiling Pt: 39.8 Melting Pt: -95 Density: 1.325 Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1 III Merck Index: 14,6063 |
|
Specification Test Results
Evaporation residue | 3 ppm |
Chlorinated pesticides | 5 ppt |
H2O | 0.0002 |
Free halogens | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...