2-Propanol ≥99.5% semiconductor grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
19397-AP
19397-K2
19397-K7
19397-M1
19397-M6
CAAA19397-APEA
76.95
CAD
CAAA19397-AP
CAAA19397-K2
CAAA19397-K7
CAAA19397-M1
CAAA19397-M6
2-Propanol ≥99.5% semiconductor grade
2-Propanol
MDL: MFCD00011674
Beilstein Registry No.: 635639
Purity: 99.5% min
CAS No.: 67-63-0
Molecular Formula: (CH3)2CHOH
Formula Weight: 60.10
Flash Point: 12°(53°F)
Physical Form: Liquid
Density: 0.785
Refractive Index: 1.3770
Odor: Alcohol-like
Melting Point: -90°
Boiling Point: 82-83°
Beilstein Reference: 635639
Merck Reference: 14,5208
UN No.: 1219
MDL No.: MFCD00011674
Formula:
(CH₃)₂CHOH Boiling Pt: 82…83 °C Melting Pt: –90 °C Density: 0.785 Flash Pt: 12 °C (53 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II REACH: 01-2119457558-25 Merck Index: 14,5208 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...