PolarSafe™ Polypropylene Storage Boxes, 15 and 50 ml, Argos Technologies
Supplier: Antyila Scientific
04396-55 04396-56
89128-208CS 821.14 CAD
89128-208 89128-210
PolarSafe™ Polypropylene Storage Boxes, 15 and 50 ml, Argos Technologies
Boxes Cryoboxes
Polypropylene freezer boxes accommodate 15 & 50mL centrifuge tubes for long term storage; an excellent alternative to cardboard storage boxes
Boxes can be stored to -90°C (130°F) and are autoclavable to 122°C (251.6°F). Accommodate most 4" stainless steel freezer racks.
Clear lid and easy-to-read numerical grid allows for tube reference and identification. White top portion allows for easy labeling or marking.