Sodium chloride, high purity
Supplier: VWR
Synonyms:
Halite
, Sea salt
, NaCl
, Saline
VWRVX190-1KG
VWRVX190-5KG
97063-366CS
1446.06
CAD
97063-368
97063-366
Sodium chloride, high purity
Sodium chloride
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 °C (1013 hPa) Melting Pt: 801 °C Density: 2.16 g/cm³ (20 °C) |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 Merck Index: 14,08599 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...