D-(+)-Sucrose ≥99.9%, Proteomics Grade DNase-, RNase-free
Supplier: VWR
M117-1KG
M117-500G
97063-788CS
467.71
CAD
97063-788
97063-790
D-(+)-Sucrose ≥99.9%, Proteomics Grade DNase-, RNase-free
D(+)-Sucrose
Formula:
C₁₂H₂₂O₁₁ Melting Pt: 169…170 °C Density: 1.29 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006626 CAS Number: 57-50-1 EINECS: 200-334-9 Merck Index: 13,08966 |
|
Specification Test Results
Chloride |
|
DNase | none detected |
Glucose |
|
Heavy Metals (as Pb) |
|
Appearance | Fine white crystals |
Insolubles |
|
Invert Sugar |
|
Iron |
|
Loss on Drying |
|
Protease | none |
Purity |
|
RNase | none detected |
Solubility (10%, Water) | Pass |
Specific Rotation | 66.3°-66.8° |
Sulfate |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...