Guanidinium hydrochloride, Proteomics Grade

Supplier: VWR
Warning

M110-500G
97063-764EA 159.3 CAD
97063-764 97063-766
Guanidinium hydrochloride, Proteomics Grade
Guanidine hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
Melting Pt: 180 °C…185 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@230nm (6M, Water) 0.15
Abs.@260nm (6M, Water) 0.03
Arsenic 0.0005 %
DNase none detected
Iron 0.0005 %
Lead 0.0005 %
Melting Point 181°C - 187°C
Protease none detected
Purity 99.5 %
RNase none
Solubility (6M, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR