Ammonium sulfate, Proteomics Grade

Supplier: VWR
Warning

Synonyms: Diammonium sulfateSulphuric acid diammonium saltsulfuric acid diammonium saltDiammonium sulphateMascagnite

VWRVM105-1KG VWRVM105-5KG
97063-746CS 305.18 CAD
97063-746 97063-748
Ammonium sulfate, Proteomics Grade
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 280 °C
Density: 1.78 g/cm³ (20 °C)
MDL Number: MFCD00003391
CAS Number: 7783-20-2
Merck Index: 13,00559

Order Now

Specification Test Results

Arsenic 0.0001%
Chloride 0.0005 %
DNase none detected
Heavy Metals (as Pb) 0.0002 %
Insolubles 0.005 %
Iron 0.0005 %
Magnesium Test Pass
pH (5%, Water) @25°C 5.0 - 6.0
Phosphate 0.0002 %
Protease none
Purity (Dry Basis) 99.5 %
Residue on Ignition 0.005 %
RNase none detected
Selenium = 0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR