Glycine, Proteomics Grade
Supplier: VWR
M103-5KG
M103-1KG
97063-738EA
646.31
CAD
97063-738
97063-736
Glycine, Proteomics Grade
Glycine
Formula:
NH₂CH₂COOH Boiling Pt: 240 °C Melting Pt: 83 °C Density: 1.64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...