Leupeptin
Supplier: VWR
J580-5MG
J580-25MG
97063-234EA
152.77
CAD
97063-234
97064-236
Leupeptin
Proteins and Peptides
A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin
- Reversible inhibitor
- Ultra pure grade
Inhibits trypsin, plasmin, papain, kallikrein, thrombin and cathepsin A and B.