Mineral oil, white (petroleum) light, high purity

Supplier: VWR
Warning

J217-100ML J217-500ML J217-1L
97064-128EA 51.19 CAD
97064-128 97064-130 490015-808
Mineral oil, white (petroleum) light, high purity
White mineral oil (petroleum)
Boiling Pt: 260…426.7 °C
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5
EINECS: 232-455-8

Order Now

Specification Test Results

Specific Gravity 0.838 - 0.854
Viscosity @ 40°C 14.2 cS - 17 cS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR