β-(N-Morpholino)ethanesulfonic acid (MES), anhydrous ≥99%, high purity suitable for buffer solutions
Supplier: VWR
E183-250G
E183-500G
E183-100G
97062-660CS
6970.33
CAD
97062-660
97062-662
97062-658
β-(N-Morpholino)ethanesulfonic acid (MES), anhydrous ≥99%, high purity suitable for buffer solutions
β-(N-Morpholino)ethanesulphonic acid (MES)
Formula:
C₆H₁₃NO₄S Melting Pt: 316 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006181 CAS Number: 4432-31-9 EINECS: 224-632-3 Merck Index: 13,05929 |
Specification Test Results
Abs.@290nm (20%, Water) (w/w) |
|
Heavy Metals |
|
pKa (20°C) | 5.9 - 6.3 |
Purity |
|
Solubility (10%, Water) | Pass |
Water (KF) |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...