Tricine, Ultrapure

Supplier: VWR

Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine

E170-500G E170-100G
97062-646CS 4258.51 CAD
97062-646 97062-642
Tricine, Ultrapure
Tricine

Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH.

Formula: (HOCH₂)₃CNHCH₂CO₂H
Melting Pt: 187 °C
Density: 1.523 g/cm³ (19.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS Number: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Order Now

Specification Test Results

DNase none detected
Lead = 0.001 %
Melting Point 182°C - 188°C
pKa @25°C 8.1 - 8.2
Protease none detected
Purity = 99 %
RNase none detected
Solubility (10%, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR