β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid, Ultrapure

Supplier: VWR
Warning

Synonyms: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate, 2-Morpholinoethanesulfonic acid hydrate, 2-(N-Morpholino)ethanesulfonic acid monohydrate

E169-100G E169-250G E169-500G
97062-636CS 2894.81 CAD
97062-636 97062-638 97062-640
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid, Ultrapure
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate

Biological buffers are useful for cell culture in vitro, enzyme assays, and some electrophoretic applications at physiological pH.

Formula: C₆H₁₃NO₄S·1H₂O
MW: 213.25 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929

Order Now

Specification Test Results

Abs.@260nm (1M, Water) 0.02
Abs.@280nm (1M, Water) 0.02
DNase none detected
Appearance White crystalline powder
Moisture (KF) 10 %
pKa @25°C 5.9 - 6.3
Protease none detected
Purity (Dry Basis,Titration) 99 %
RNase none detected
Solubility (1M, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR