Potassium hydrogen carbonate, reagent

Supplier: VWR

Synonyms: Potassium bicarbonate

0889-1KG 0889-500G
97062-492CS 328.51 CAD
97062-492 97062-496
Potassium hydrogen carbonate, reagent
Potassium hydrogen carbonate
Formula: KHCO₃
MW: 100.12 g/mol
Melting Pt: 292
Density: 2.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR