Potassium carbonate, anhydrous ACS

Supplier: VWR
Warning

Synonyms: Potash

0879-1KG 0879-2.5KG 0879-500G
97062-470CS 508.25 CAD
97062-470 97062-472 97062-474
Potassium carbonate, anhydrous ACS
Potassium carbonate
Formula: K₂CO₃
Melting Pt: 891 °C
Density: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium 0.005%
Chloride = 0.003%
Heavy Metals = 0.0005%
Insolubles = 0.01%
Iron = 0.0005%
Magnesium = 0.002%
Phosphate = 0.001%
Purity = 99%
Silica = 0.005%
Sodium = 0.02%
Sulfur Compounds = 0.004%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR