Sodium sulfate, anhydrous ACS

Supplier: VWR

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

0836-500G 0836-1KG 0836-2.5KG
97062-438CS 1171.87 CAD
97062-438 97062-434 97062-436
Sodium sulfate, anhydrous ACS
Sodium sulphate
Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 888 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium 0.01%
Chloride = 0.001%
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.01%
Iron = 0.001%
Loss on Ignition = 0.5%
Magnesium = 0.005%
Nitrogen Compounds = 0.0005%
pH (5%, Water) @25°C 5.2 - 9.2
Phosphate = 0.001%
Potassium = 0.01%
Purity = 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR