Potassium dihydrogen phosphate, anhydrous ACS

Supplier: VWR

0781-2.5KG 0781-12KG 0781-1KG 0781-500G
97062-348CS 1106.46 CAD
97062-348 97062-344 97062-346 97062-350
Potassium dihydrogen phosphate, anhydrous ACS
Potassium dihydrogen phosphate
Formula: KH₂PO₄
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 253 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Order Now

Specification Test Results

Chloride 0.001%
Heavy Metals = 0.001%
Insolubles = 0.01%
Iron = 0.002%
Loss on Drying = 0.2%
Nitrogen Compounds = 0.001%
pH (5%, Water) @25°C 4.1 - 4.5
Purity = 99%
Sodium = 0.005%
Sulfate = 0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR