Polysorbate 20, Reagent Grade

Supplier: VWR

Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated

0777-1L 0777-4L
97062-332EA 163.04 CAD
97062-332 97062-334
Polysorbate 20, Reagent Grade
Polysorbate 20

Non-ionic detergent for solubilizing membrane proteins without altering biological activity.

Formula: C₅₈H₁₁₄O₂₆
Boiling Pt: > 100 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR