TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Reagent Grade
Supplier: VWR
|
Danger
|
0747-4L
0747-1L
97062-308EA
304.92
CAD
97062-308
97062-306
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Reagent Grade
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether)
Non-ionic - containing no charge (polyoxyethylenes or alkyl glucosides). Non-ionic detergents are suited for solubilizing embrane proteins without altering biological activity. These non-denaturing, gentler detergents are ideal for studying the conformati
Boiling Pt:
∼150 °C (1013 hPa) Density: 1.055 g/cm³ (20 °C) Flash Pt: >93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128254 CAS Number: 9036-19-5 UN: 3082 ADR: 9 III |
Specification Test Results
Cloud Point (1%, Water) | 22 °C - 28 °C |
Color (APHA) |
|
Index of Refraction | 1.48 - 1.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...