, anhydrous ACS

Supplier: VWR

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

0705-500G 0705-12KG 0705-2.5KG
97062-234CS 1744.81 CAD
97062-234 97062-230 97062-232
, anhydrous ACS
di-Potassium hydrogen phosphate
Formula: K₂HPO₄
Melting Pt: 340 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride 0.003%
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.01%
Iron = 0.001%
Loss on Drying = 1%
Nitrogen Compounds = 0.001%
pH (5%, Water) @25°C 8.5 - 9.6
Purity = 98%
Sodium = 0.05%
Sulfate = 0.005%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR