Potassium acetate ACS

Supplier: VWR

0698-2.5KG 0698-1KG
97062-214CS 1504.25 CAD
97062-214 97062-212
Potassium acetate ACS
Potassium acetate
Formula: CH₃COOK
Melting Pt: 304 °C
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium 0.005%
Chloride = 0.003%
Heavy Metals = 0.0005%
Insolubles = 0.005%
Iron = 0.0005%
Magnesium = 0.002%
pH (5%, Water) @25°C 6.5 - 9
Phosphate = 0.001%
Purity = 99%
Sodium = 0.03%
Sulfate = 0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR